Learn More
ATG9A Mouse anti-Human, Unconjugated, Clone: 1D12E11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67096-1-IG-20UL
 View more versions of this product
Product Code. 16849525
  • 1710.33 NOK / 20µL
Description
The ATG9A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATG9A. This antibody recognizes human antigen. The ATG9A antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
ATG9A
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1D12E11
Unconjugated
20 μL
79065
IgG1