missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG4D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16924-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some maligt tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases.Specifications
ATG4D | |
Polyclonal | |
Unconjugated | |
ATG4D | |
APG4 D, APG4D, ATG4D, AUTL4, Autophagin 4, Cysteine protease ATG4D | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
235040, 686505, 84971 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86TL0, Q8BGV9 | |
ATG4D | |
ATG4D Fusion Protein Ag10307 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |