missing translation for 'onlineSavingsMsg'
Learn More

Ataxin 2 Rabbit anti-Human, Mouse, Rat, Biotin, Polyclonal, Proteintech

Product Code. 17761239 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
Unit Size:
20µL
Product Code. Quantity unitSize
17761239 20 μL 20µL
1 options
This item is not returnable. View return policy

Product Code. 17761239

Brand: Proteintech Biotin2177620UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ataxin 2 Polyclonal antibody specifically detects Ataxin 2 in Human, Mouse, Rat samples. It is validated for IHC

Specifications

Antigen Ataxin 2
Applications Immunohistochemistry
Classification Polyclonal
Concentration 470 μg/mL
Conjugate Biotin
Formulation PBS with 50% Glycerol, 0.05% Proclin300, pH 7.3.
Gene Accession No. BC000614
Host Species Rabbit
Purification Method Antigen affinity purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 6311
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.