missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ataxin 2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15712121 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
15712121 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 15712121

Brand: Invitrogen™ PA554183

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EGHSINTREN KYIPPGQRNR EVISWGSGRQ NSPRMGQPGS GSMPSRSTSH TSDFNPNSGS DQRVVNGGVP WPSPCPSPSS RPPSRYQSGP NSLPPRAATP TRPPSRPPSR PSRPPSHPSA HGSPAPVSTM PKRMSSE Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.

The autosomal dominant cerebellar ataxias (ADCA) are a heterogeneous group of neurodegenerative disorders characterized by progressive degeneration of the cerebellum, brain stem and spinal cord. Clinically, ADCA has been divided into three groups: ADCA types I-III. Defects in this gene are the cause of spinocerebellar ataxia type 2 (SCA2). SCA2 belongs to the autosomal dominant cerebellar ataxias type I (ADCA I) which are characterized by cerebellar ataxia in combination with additional clinical features like optic atrophy, ophthalmoplegia, bulbar and extrapyramidal signs, peripheral neuropathy and dementia. SCA2 is caused by expansion of a CAG repeat in the coding region of this gene. Longer expansions result in earlier onset of the disease. Alternatively spliced transcript variants encoding different isoforms have been identified but their full length sequence has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ataxin 2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Atxn2
Gene Accession No. Q99700
Gene Alias 9630045M23Rik; ASL13; ataxin 2; ataxin-2; ATX2; Atxn2; AW544490; FLJ46772; SCA2; spinocerebellar ataxia 2 (olivopontocerebellar ataxia 2, autosomal dominant, ataxin 2); spinocerebellar ataxia 2 homolog; spinocerebellar ataxia type 2 protein; Spinocerebellar ataxia type 2 protein homolog; TNRC13; Trinucleotide repeat-containing gene 13 protein
Gene Symbols Atxn2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Ataxin 2. Recombinant protein control fragment (Product #RP-91710).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6311
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.