Learn More
ATAGO™ Digital Refractometer PR-201α
Description
Allows you to measure Brix values of jams, concentrated juices, coolants and other samples which cannot be measured with the PR-101μ.
Specifications
Specifications
| Type | Refractometer |
| Scale Range | 0.0 to 60.0 Brix |
| Model | PR-201α |
| Accuracy | ± 0.1% |
| Resolution | Min. Indication 0.1% |
| Scale Type | Brix |
| Ambient Temperature (Metric) | 10°C to 40°C |
| Temperature Compensation | 10°C to 40°C |