missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATAD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16610-1-AP-150UL
This item is not returnable.
View return policy
Description
ATAD3B is a member of the AAA ATPase family, a family of proteins that catalyze ATP into ADP and are involved in several cellular functions such as cell-cycle regulation, protein proteolysis and transport. The AAA ATPase family is characterized by a highly conserved AAA motif containing Walker homology sequences and imparting ATPase activity. Mitochondrial membrane proteins ATAD3A/B contribute to the stabilization of nucleoids and may participate in the transformation pathway and the chemosensitivity of oligodendrogliomas. The gene encoding ATAD3A/B/C maps to human chromosome 1 and has been suggested to be an anti-apoptotic marker.Specifications
| ATAD3 | |
| Polyclonal | |
| Unconjugated | |
| ATAD3B | |
| AAA TOB3, ATAD3B, KIAA1273, TOB3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108888, 298682, 83858 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5T9A4 | |
| Atad3a, ATAD3B | |
| ATAD3 Fusion Protein Ag9836 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction