Learn More
Asunaprevir, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
Description
Asunaprevir (BMS-650032) is a potent and orally bioavailable hepatitis C virus (HCV) NS3 protease inhibitor, with IC50 of 0.2 nM-3.5 nM. Asunaprevir inhibits SARS-CoV-2 3CLpro activity.
Specifications
Specifications
| Chemical Name or Material | Asunaprevir |
| CAS | 630420-16-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C35H46ClN5O9S |
| Quantity | 10 mg |
| Synonym | BMS-650032 |
| Solubility Information | DMSO : ≥ 100 mg/mL (133.64 mM) ∣Ethanol : 20 mg/mL (26.73 mM; Need ultrasonic) |
| SMILES | ClC1=CC=C2C(C(O[C@H]3CN(C([C@@H](NC(OC(C)(C)C)=O)C(C)(C)C)=O)[C@H](C(N[C@@]4(C(NS(C5CC5)(=O)=O)=O)[C@H](C=C)C4)=O)C3)=NC=C2OC)=C1 |
| Molecular Weight (g/mol) | 748.29 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.