Learn More
ASTX660, MedChemExpress
Product Code. 16493026
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. 16493026
Brand: Medchem Express HY109565
MedChemExpress ASTX660 is an orally bioavailable dual antagonist of cellular inhibitor of apoptosis protein (cIAP) and X-linked inhibitor of apoptosis protein (XIAP).
ASTX660 is an orally bioavailable dual antagonist of cellular inhibitor of apoptosis protein (cIAP) and X-linked inhibitor of apoptosis protein (XIAP).
Specifications
Chemical Name or Material ASTX660
CAS 1799328-86-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H42FN5O3
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (185.29 mM; Need ultrasonic)
SMILES C[C@H]1N(C[C@@H]2N(CC(N3CC(C)(C)C4=NC(CO)=C(CC5=CC=C(F)C=C5)C=C43)=O)C[C@@H](C)NC2)CCOC1
Molecular Weight (g/mol) 539.68
Formula Weight 539.68
Percent Purity 98.99%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.