Learn More
ASTM™ D6584 Standard Solution 2, Honeywell Fluka™
Product Code. 15940192 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
15940192 1 mL 1mL
1 options
Product Code. 15940192
Brand: Honeywell Fluka R13881ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
in pyridne, analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Packaging Glass Bottle
CAS 110-86-1, 111-03-5
Density 0.98 g/cm3
Flash Point 20°C
Physical Form Liquid
Boiling Point 115°C
Industry Type Biofuels
Quantity 1 mL
Grade Analytical Standard