Learn More
ASTM™ D6584 Monoolein solution, Honeywell Fluka™
Product Code. 15929701 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
3 mL
Unit Size:
3mL
Product Code. Quantity unitSize
15929701 3 mL 3mL
1 options
Product Code. 15929701
Brand: Honeywell Fluka R13813ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
5000 μg/mL in pyridine, analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Packaging Glass Bottle
CAS 110-86-1, 111-03-5
Density 0.98 g/cm3
Flash Point 20°C
Physical Form Liquid
Boiling Point 115°C
Industry Type Biofuels
Quantity 3 mL
Grade Analytical Standard