Learn More
ASTM™ D6584 1,3-Diolein solution, Honeywell Fluka™
Product Code. 15939701 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
Product Code. Quantity unitSize
15939701 2 mL 2mL
1 options
Product Code. 15939701
Brand: Honeywell Fluka R13822ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
5000 μg/mL in pyridine, analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Packaging Glass Bottle
CAS 2465-32-9, 110-86-1
Density 0.98 g/cm3
Flash Point 20°C
Physical Form Liquid
Boiling Point 115°C
Industry Type Biofuels
Quantity 2 mL
Grade Analytical Standard