missing translation for 'onlineSavingsMsg'
Learn More

ASTM D5453 Curve 1, S @ 0.5-10ppm in p-xylene, VHG™

Product Code. 17941726
Change view
Click to view available options
Quantity:
6 x 2 mL
Unit Size:
12mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17941726 6 x 2 mL 12mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17941726 Supplier VHG Labs Supplier No. ALKKTSPXV6CRV1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ASTM D5453 Curve 1, S @ 0.5-10ppm in p-xylene
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfur
CAS 7704-34-9
Analyte or Component Names Sulfur
Shipping Condition Room Temperature
Molecular Formula S
Concentration 0.5-10 ppm
InChI Formula InChI=1S/S
SMILES [S]
IUPAC Name sulfur
Solvent or Matrix p-xylene
Molecular Weight (g/mol) 32.065
Quantity 6 x 2 mL
Formula Weight 31.972 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.