missing translation for 'onlineSavingsMsg'
Learn More

AST 487, MedChemExpress

Product Code. p-200097838
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488867

Brand: Medchem Express HY15002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AST 487 is a RET kinase inhibitor with IC50 of 880 nM, inhibits RET autophosphorylation and activation of downstream effectors, also inhibits Flt-3 with IC50 of 520 nM.

AST 487 is a RET kinase inhibitor with IC50 of 880 nM, inhibits RET autophosphorylation and activation of downstream effectors, also inhibits Flt-3 with IC50 of 520 nM.

Specifications

Chemical Name or Material AST 487
CAS 630124-46-8
Color White
Purity Grade Notes Research
Molecular Formula C26H30F3N7O2
Quantity 5 mg
Synonym NVP-AST 487
Solubility Information DMSO : ≥ 100 mg/mL (188.84 mM)
SMILES O=C(NC1=CC=C(OC2=NC=NC(NC)=C2)C=C1)NC3=CC=C(CN4CCN(CC)CC4)C(C(F)(F)F)=C3
Molecular Weight (g/mol) 529.56
Formula Weight 529.56
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.