Learn More
Karl Hecht™ Assistent™ Glass Vials
Product Code. 10227391
Change view
Click to view available options
Volume (Metric):
8 mL
20 mL
Height (Metric):
34 mm
60 mm
Diameter (Metric) Outer:
24 mm
26 mm
Unit Size:
Pack of 130
Pack of 154
Product Code. Volume (Metric) Height (Metric) Diameter (Metric) Outer unitSize
10227391 20 mL 60 mm 26 mm Pack of 130
10145291 8 mL 34 mm 24 mm Pack of 154
2 options
Product Code. 10227391
Brand: Karl Hecht™ 42783024
Only null left
Add to basket
Add to basket
Suitable for media, diagnostic, storage, display and sample collection applications. Karl Hecht™ Assistent™ Glass Vials come with a plastic snap-cap for secure closure.
  • Clear glass
  • With rolled rim
Specifications
Diameter (Metric) Outer 26 mm
Volume (Metric) 20 mL
Closure Type Snap Cap
Height (Metric) 60 mm