Learn More
Hecht Karl™ Assistent™ Borosilicate Weighing Bottles
Click to view available options
Capacity (Metric):
10 mL
15 mL
30 mL
45 mL
70 mL
80 mL
110 mL
Unit Size:
Each
Specifications
Specifications
| Capacity (Metric) | 110 mL |
| Includes | Interchangeable NS cover and frosted label |
| Material | Borosilicate Glass |
| Product Type | Weighing Bottle |
| Closure Type | NS Cover |