Learn More
Thermo Scientific™ Assembled Caps and Septa for 8-425 Standard-Opening Vials
Click to view available options
Color:
Black
Blue
Natural
Red
White
Yellow
Unit Size:
Pack of 100
Description
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials for a wide range of applications.
- Open top caps can be used with any 8mm septa
- Phenolic caps are suitable for autoclaving and low temperature applications
- Polypropylene caps are chemically inert and suitable for most chromatography applications
- Kim-Lok™ cap/septum assemblies feature a molded centering ring to eliminate septum movement during injection
- PTFE/red rubber septa are economical for many GC applications
- PTFE/silicone septa are most popular for HPLC and IC applications
- Pre-slit septa are easier to pierce with thin-gauge needles
Specifications
Specifications
| Description | PP, Blue; Kim-Lok Red PTFE/White Silicone septum |
| Color | Blue |
| Material | PTFE, Silicone, Polypropylene |
| Septum | Kim-Lok Red PTFE/White Silicone |
| Type | Kim-Lok Cap with Flange |
| Closure Size | 8-425 |
| For Use With (Equipment) | 8-425 Standard Opening Vials |
Product Content Correction
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto