Learn More
Hirschmann™ Aspirette™ Vacuum Tweezers
Product Code. 10454861
Change view
Click to view available options
:
230 Volt; Euro plug
Product Code.
10454861 230 Volt; Euro plug
1 options
Product Code. 10454861
Brand: Hirschmann™ 9901500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Vacuum tweezers for picking up and positioning diverse plane objects up to a maximum piece weight of 70 g.
  • Delivery complete with 1 power supply unit and charger
  • Includes 3 suction nozzles and suction cups with 3.2, 6 and 10 mm diameters
  • 230 Volt, Euro plug
Specifications
Electrical Requirements 230 V, Euro plug
For Use With (Application) Picking up, positioning plane objects
Type Vacuum Tweezers