missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ASPH Monoclonal Antibody (C13), Invitrogen™

Product Code. 17604406
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
Product Code. Quantity unitSize
17604406 20 μL 20µL
17694396 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17604406

Brand: Invitrogen™ MA543946

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ASPH is thought to play an important role in calcium homeostasis. Alternative splicing of this gene results in five transcript variants which vary in protein translation, the coding of catalytic domains, and tissue expression. Variation among these transcripts impacts their functions which involve roles in the calcium storage and release process in the endoplasmic and sarcoplasmic reticulum as well as hydroxylation of aspartic acid and asparagine in epidermal growth factor-like domains of various proteins. This gene is thought to play an important role in calcium homeostasis. Alternative splicing of this gene results in five transcript variants which vary in protein translation, the coding of catalytic domains, and tissue expression. Variation among these transcripts impacts their functions which involve roles in the calcium storage and release process in the endoplasmic and sarcoplasmic reticulum as well as hydroxylation of aspartic acid and asparagine in epidermal growth factor-like domains of various proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASPH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Asph
Gene Accession No. Q12797
Gene Alias 2310005F16Rik; 3110001L23Rik; A beta H-J-J; AAH; AI848629; ASP beta-hydroxylase; Aspartate beta-hydroxylase; aspartate-beta-hydroxylase; aspartyl (asparaginyl) beta hydroxylase; aspartyl beta-hydroxylase; aspartyl/asparaginyl beta-hydroxylase; aspartyl/asparaginyl beta-hydroxylase-like; aspartyl/asparaginyl-beta-hydroxylase; ASPH; AW261690; AW561901; BAH; C79816; calsequestrin-binding protein; cardiac junctin; CASQ2BP1; cI-37; FDLAB; HAAH; humbug; JCTN; jumbug; junctate; junctin; junctional sarcoplasmic reticulum protein; peptide-aspartate beta-dioxygenase; Unknown (protein for MGC:137539)
Gene Symbols Asph
Host Species Mouse
Immunogen Leu345-Ile758 (Accession Q12797), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 444
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.