Learn More
ASK1 Mouse anti-Human, Unconjugated, Clone: 1C3A10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67072-1-IG-20UL
 View more versions of this product
Product Code. 16849485
  • 1710.33 NOK / 20µL
Description
The ASK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ASK1. This antibody recognizes human, mouse, rat antigen. The ASK1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
ASK1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1C3A10
Unconjugated
20 μL
4217
IgG1