missing translation for 'onlineSavingsMsg'
Learn More

Asivatrep, MedChemExpress

Product Code. 16445976
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. Quantity unitSize
16445976 100 mg 100mg
16485338 10 mg 10mg
16458057 5 mg 5mg
16451537 50 mg 50mg
16441299 1 mg 1mg
5 options
This item is not returnable. View return policy

Product Code. 16445976

Brand: Medchem Express HY12777

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Asivatrep (PAC-14028) is a potent and selective transient receptor potential vanilloid type I (TRPV1) antagonist.

Asivatrep (PAC-14028) is a potent and selective transient receptor potential vanilloid type I (TRPV1) antagonist.

Specifications

Chemical Name or Material Asivatrep
CAS 1005168-10-4
Color White
Purity Grade Notes Research
Molecular Formula C21H22F5N3O3S
Quantity 100 mg
Synonym PAC-14028
Solubility Information DMSO : 50 mg/mL (101.74 mM; Need ultrasonic)
SMILES O=C(N[C@@H](C1=CC(F)=C(NS(=O)(C)=O)C(F)=C1)C)/C=C/C2=CC=C(C(F)(F)F)N=C2CCC
Molecular Weight (g/mol) 491.47
Formula Weight 491.47
Percent Purity 95.14%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.