Learn More
Invitrogen™ ASIC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113613
Description
Immunogen sequence: LLDLLGKEED EGSHDENVST CDTMPNHSET ISHTVNVPLQ TTLGTLEEIA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
ACCN1 encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, 2 hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. The member encoded by this protein may play a role in neurotransmission. In addition, a heteromeric association between this member and ACCN3 (variant 1) has been observed to co-assemble into proton-gated channels sensitive to gadolinium.
Specifications
| ASIC2 | |
| Polyclonal | |
| Unconjugated | |
| ASIC2 | |
| ACCN; ACCN1; ACIC2; acid sensing (proton gated) ion channel 2; acid sensing ion channel 2; acid sensing ion channel subunit 2; acid-sensing (proton-gated) ion channel 2; acid-sensing ion channel 2; Amiloride-sensitive brain sodium channel; amiloride-sensitive brain sodium channel 2; Amiloride-sensitive cation channel 1, neuronal; amiloride-sensitive cation channel 1, neuronal (degenerin); Amiloride-sensitive cation channel neuronal 1; Asic2; ASIC2a; Bnac1; BNaC1a; BNC1; BNC1k; Brain sodium channel 1; degenerin; hBNaC1; Mammalian degenerin homolog; MDEG; MDEG1; MDEG2; neuronal amiloride-sensitive cation channel 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 40 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q16515 | |
| ASIC2 | |
| Synthetic peptide directed towards the middle region of human ACCN1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.