Learn More
ASF1B Monoclonal Antibody (OTI4A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA813137
Encompass_Preferred
 View more versions of this product
Product Code. 17577854
  • 6295.38 NOK / 100µL
Description
ASF1B Monoclonal antibody specifically detects ASF1B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
ASF1B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9DAP7, Q9NVP2
Mouse
Protein A/G
RUO
304648, 55723, 66929
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI4A4
Unconjugated
ASF1B
1700003K02Rik; AA409591; anti-silencing function 1B histone chaperone; anti-silencing function protein 1 homolog B; ASF1 anti-silencing function 1 homolog B; Asf1b; CCG1-interacting factor A-II; CIA-II; hAsf1; hAsf1b; hCIA-II; histone chaperone ASF1B; mCIA-II
Full length human recombinant protein of human ASF1B (NP_060624) produced in E.coli.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.