missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASF1B Monoclonal Antibody (OTI1E9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
2816.72 NOK - 6295.38 NOK
Specifications
| Antigen | ASF1B |
|---|---|
| Clone | OTI1E9 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17547844
|
Origene Technologies
TA813129S |
30 μL |
2816.72 NOK
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17527844
|
Origene Technologies
TA813129 |
100 μL |
6295.38 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASF1B Monoclonal antibody specifically detects ASF1B in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ASF1B | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| 1700003K02Rik; AA409591; anti-silencing function 1B histone chaperone; anti-silencing function protein 1 homolog B; ASF1 anti-silencing function 1 homolog B; Asf1b; CCG1-interacting factor A-II; CIA-II; hAsf1; hAsf1b; hCIA-II; histone chaperone ASF1B; mCIA-II | |
| Full length human recombinant protein of human ASF1B (NP_060624) produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| ASF1B |
| OTI1E9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q9DAP7, Q9NVP2 | |
| 304648, 55723, 66929 | |
| IgG2a | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title