missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ASB5 Polyclonal antibody specifically detects ASB5 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ASB5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | ankyrin repeat and SOCS box containing 5, ankyrin repeat and SOCS box protein 5, ankyrin repeat and SOCS box-containing 5, ASB-5, SOCS box protein ASB-5 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 130-329 of human ASB5 (NP_543150.1). NAITIDGVTPLFNACSQGSPSCAELLLEYGAKAQLESCLPSPTHEAASKGHHECLDILISWGIDVDQEIPHLGTPLYVACMSQQFHCIWKLLYAGADVQKGKYWDTPLHAAAQQSSTEIVNLLLEFGADINAKNTELLRPIDVATSSSMVERILLQHEATPSSLYQLCRLCIRSYIGKPRLHLIPQLQLPTLLKNFLQYR |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?