missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASB13 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25616-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing family of proteins. They contain ankyrin repeat sequence and a SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length sequences are not known.Specifications
| ASB13 | |
| Polyclonal | |
| Unconjugated | |
| ASB13 | |
| ASB 13, ASB13 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 142688, 79754 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8VBX0, Q8WXK3 | |
| ASB13 | |
| ASB13 Fusion Protein Ag22533 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction