Learn More
Asatone, MedChemExpress
Product Code. 16492599
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. 16492599
Brand: Medchem Express HYN6826
MedChemExpress Asatone is an active component isolated from Radix et Rhizoma Asari, with anti-inflammatory effect via activation of NF-κB and donwn regulation of p-MAPK (ERK, JNK and p38) pathways.
Asatone is an active component isolated from Radix et Rhizoma Asari, with anti-inflammatory effect via activation of NF-κB and donwn regulation of p-MAPK (ERK, JNK and p38) pathways.
Specifications
Chemical Name or Material Asatone
CAS 38451-63-7
Color White
Purity Grade Notes Research
Molecular Formula C24H32O8
Quantity 1 mg
Solubility Information DMSO : 100 mg/mL (222.96 mM; Need ultrasonic)
SMILES O=C1C(OC)(OC)[C@@]2([H])[C@@](C(OC)(OC)C3=O)([H])C(CC=C)=C[C@@]3(OC)[C@@]2(CC=C)C=C1OC
Molecular Weight (g/mol) 448.51
Formula Weight 448.51
Percent Purity 99.88%
Grade Research
Physical Form Crystals
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.