Learn More
Thermo Scientific™ ASAPath™ COVID-19 Antigen Rapid Test (CE-IVD)
Product Code. 17286053 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
10 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17286053 1 Kit 10 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17286053 Supplier Thermo Scientific™ Supplier No. 10028730
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Obtain COVID-19 test results fast with the easy-to-use, disposable Thermo Scientific ASAPath COVID-19 Antigen Rapid Test.
Obtain COVID-19 test results within 30 minutes with the fast, easy-to-use, disposable Thermo Scientific™ ASAPath™ COVID-19 Antigen Rapid Test. This test is a qualitative lateral flow immunoassay intended to detect the nucleocapsid protein from the SARS-CoV-2 virus, which causes COVID-19, in nasal swab samples from symptomatic individuals who are suspected of COVID-19 by a healthcare professional, or individuals with or without symptoms or other epidemiological reasons to suspect COVID-19 infection.
  • Rapid Antigen Test for COVID-19
  • Detect nasal swab sample
  • Kit contains 10 sets of tests
  • Easy nasal swab sample collection
  • Test can be done by a single person
  • Results within 30 minutes
  • Easy-to-read results
  • No special reader necessary
  • Antibody pool targeting several virus protein regions for variant detection
  • Single use, disposable device
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room Temperature
Description ASAPath™ COVID-19 Antigen Rapid Test (CE-IVD)
Includes One kit contains ten (10) Antigen Rapid Tests, each of which includes cassette, swab, extraction tube, dropper cap, and tube stand
Quantity 1 Kit
compliance-icons
Product Identifier
  • X10 ASAPath Covid-19 Antigen Rapid Test
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Hexadecan-1-yl(trimethyl)ammonium chloride
Product is CE Marked but not 510(k) cleared nor authorized for Emergency Use in US and not yet available for sale in the U.S. Availability of the product in each country depends on local regulatory marketing authorization status.