missing translation for 'onlineSavingsMsg'
Learn More

ASAH3 Antibody, Novus Biologicals™

Product Code. p-7109008 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18297731 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18297731 Supplier Novus Biologicals Supplier No. NBP174125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASAH3 Polyclonal specifically detects ASAH3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASAH3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8TDN7
Gene Alias Acylsphingosine deacylase 3, Alkaline CDase 1, alkaline ceramidase 1, AlkCDase 1, ALKCDase1, ASAH3, EC 3.5.1.23, MGC138327, MGC138329, N-acylsphingosine amidohydrolase (alkaline ceramidase) 3, N-acylsphingosine amidohydrolase 3
Gene Symbols ACER1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to the C terminal of ACER1. Immunizing peptide sequence ITFPYGMVTMALVDANYEMPGETLKVRYWPRDSWPVGLPYVEIRGDDKDC.
Molecular Weight of Antigen 31 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125981
Test Specificity Expected identity based on immunogen sequence: Human: 100%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.