Learn More
RETSCH AS 300 Comfort Clamping Devices
Product Code. 10437342 Shop All Retsch Products
Click to view available options
For Use With (Application):
Dry Sieving
Wet Sieving
Unit Size:
Each
Product Code. 10437342
Brand: RETSCH 32.662.0014
Only null left
Add to basket
Add to basket
Use these clamping devices with AS 300 Sieve Shakers. Retsch™ AS 300 Comfort Clamping Devices feature an ergonomic design for easy operation.
  • For use with AS 300 Sieve Shakers
Specifications
Product Type Clamping Devices
Diameter (Metric) 315 mm
For Use With (Application) Wet Sieving
For Use With (Equipment) Analytical sieve shaker AS 300