Learn More
AS-252424, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | (5Z)-5-[(5-(4-Fluoro-2-hydroxyphenyl)-2-furanyl)methylene]-2,4-thiazolidinedione |
| CAS | 900515-16-4 |
| Molecular Formula | C14H8FNO4S |
| Quantity | 25 mg |
| Synonym | AS-252424, (5Z)-5-[[5-(4-Fluoro-2-hydroxyphenyl)-2-furanyl]methylene]-2,4-thiazolidinedione |
| InChI Formula | InChI=1S/C14H8FNO4S/c15-7-1-3-9(10(17)5-7)11-4-2-8(20-11)6-12-13(18)16-14(19)21-12/h1-6,17H,(H,16,18,19)/b12-6- |
| SMILES | Oc1cc(F)ccc1c2oc(\C=C\3/SC(=O)NC3=O)cc2 |
| IUPAC Name | (5Z)-5-[[5-(4-fluoro-2-hydroxyphenyl)furan-2-yl]methylidene]-1,3-thiazolidine-2,4-dione |
| Molecular Weight (g/mol) | 305.28 |