missing translation for 'onlineSavingsMsg'
Learn More

Artemotil, MedChemExpress

Product Code. p-200097567
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16498446

Brand: Medchem Express HYB0770

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Artemotil (β-Arteether) has antimalarial activity for the treatment of chloroquine-resistant Plasmodium falciparum malaria with an IC50 of 1.61 nM. Artemotil also has central nervous system (CNS) neurotoxicity and anorectic toxicity in rats, dogs and monkeys.

Artemotil (β-Arteether) has antimalarial activity for the treatment of chloroquine-resistant Plasmodium falciparum malaria with an IC50 of 1.61 nM. Artemotil also has central nervous system (CNS) neurotoxicity and anorectic toxicity in rats, dogs and monkeys.

Specifications

Chemical Name or Material Artemotil
Physical Form Solid
CAS 75887-54-6
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H28O5
For Use With (Application) COVID-19-immunoregulation
Synonym β-Arteether (+)-Arteether Arteether
Solubility Information DMSO : 50 mg/mL (160.05 mM; Need ultrasonic)
SMILES C[C@@H]1[C@@](CC[C@@H](C)[C@]2([H])CC[C@]3(C)O4)([H])[C@@]2(OO3)[C@]4([H])O[C@@H]1OCC
Molecular Weight (g/mol) 312.4
Formula Weight 312.4
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.