Learn More
Thermo Scientific™ ART™ SolventSafe Pipette Tips
GREENER_CHOICE
Product Code. 10252431
Click to view available options
Volume (Metric):
200 μL
1000 μL
Unit Size:
Pack of 384
Pack of 400
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 10252431
Brand: Thermo Scientific™ 5479E
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Perform applications accurately and consistently with these Solventsafe pipette tips, which seal when exposed to potential contaminants.
Perform applications consistently and accurately by using Thermo Scientific™ ART™ SolventSafe Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART tips are designed to aid in pipetting for a variety of applications.
Features:
  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
Designed to aid in pipetteting for a variety of applications:
Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
TRUSTED_SUSTAINABILITY
Specifications
Description ART SolventSafe 1000E, Filtered, Sterile, Rack
Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Model ART SolventSafe 1000E
Length (English) 3.47 in.
Length (Metric) 8.83cm
Format Rack
Tip Style Micro Point
For Use With (Equipment) Universal Fit
Product Line ART