Learn More
Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, 10, bulk
Beschreibung
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.
Features:
- Sterile tips are certified free from RNase, DNase and pyrogen
- Available in sterile, nonsterile and in various packaging options
- Manufactured according to the industry's highest quality standard
Designed to aid in pipetteting for a variety of applications:
Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Spezifikation
Spezifikation
| Description | ART 10 REACH, Filtered, Non-sterile, Bulk |
| Volume (Metric) | 10 μL |
| Filtered | Filtered |
| Sterility | Non-sterile |
| Model | ART 10 REACH |
| Length (English) | 1.71 in. |
| Length (Metric) | 4.33cm |
| Format | Bulk |
| Tip Style | Extended Length |
| For Use With (Equipment) | Universal Fit |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.