Learn More
Thermo Scientific™ ART™ Barrier Reload Insert Pipette Tips
GREENER_CHOICE
Product Code. 10083064
Click to view available options
Volume (Metric):
10 μL
20 μL
100 μL
200 μL
Model:
ART 10
ART 100
ART 100E
ART 200
ART 20E
ART 20P
Tip Style:
Low Retention, Micro Point
Micro Point
Fit for Purpose:
Sterile and PCR Ready
Unit Size:
Pack of 960
Product Code. 10083064
Brand: Thermo Scientific™ 2149PRI
Only null left
Add to basket
Add to basket
Obtain accurate and consistent performance with these pipette tips, which seal when exposed to potential contaminants.
Obtain accurate and consistent performance with Thermo Scientific™ ART™ Barrier Reload Insert Pipette Tips, which seal when exposed to potential contaminants. The ART inserts are environmentally friendly, providing individually packaged blister tubs that are made out of recycled plastic, designed to be used with the reusable ART hinged racks and creating the ART Reload System. The tips from the reload inserts clip securely into the hinged racks for efficient reloading.
Features
  • Sterile tips certified free from RNase, DNase and pyrogen
  • Available in MicroPoint, extended length, low retention and SoftFit-L™ designed for use with Rainin™ LTS™ LiteTouch™ Pipettes
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 20 μL
Filtered Filtered
Sterility Sterile
Model ART 20P
Length (English) 2 in.
Length (Metric) 5.07cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Refill
Tip Style Micro Point
Green Features Fewer resources used and less waste
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile and PCR Ready