Learn More
Thermo Scientific™ ART™ Barrier Pipette Tips in Hinged Racks
GREENER_CHOICE
Product Code. 10217184
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
100 μL
200 μL
300 μL
1000 μL
Model:
ART 10
ART 10 Ultra-micro
ART 100
ART 100E
ART 200
ART 20E Ultra-micro
ART 20P
ART Barrier
Format:
Hinged Rack
Rack
Tip Style:
Low Retention
Ultra-micro
Universal Fit
Unit Size:
Pack of 768
Pack of 960
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Model Format Tip Style unitSize
10217184 10 μL ART 10 Hinged Rack Universal Fit Pack of 960
11942689 10 μL ART 10 Hinged Rack Low Retention Pack of 960
10706815 10 μL ART 10 Ultra-micro Hinged Rack Ultra-micro Pack of 960
10032874 10 μL ART 20E Ultra-micro Hinged Rack Low Retention Pack of 960
10401755 20 μL ART 20P Hinged Rack Universal Fit Pack of 960
10276944 20 μL ART 20P Hinged Rack Low Retention Pack of 960
10777385 100 μL ART 100 Hinged Rack Universal Fit Pack of 960
10746625 100 μL ART 100E Hinged Rack Universal Fit Pack of 960
12661480 100 μL ART 100E Hinged Rack Low Retention Pack of 960
10431375 200 μL ART 200 Hinged Rack Universal Fit Pack of 960
10768735 200 μL ART 200 Hinged Rack Low Retention Pack of 960
15173606 300 μL ART Barrier Rack Universal Fit Pack of 960
15193606 300 μL ART Barrier Rack Low Retention Pack of 960
10564914 1000 μL ART Barrier Hinged Rack Universal Fit Pack of 768
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 10217184 Supplier Thermo Scientific™ Supplier No. 2139HR
  • ONLINE EXCLUSIVE PRICE
    1435.72 NOK
    1268.15 NOK / Pack of 960
    Save 167.57 NOK
    11% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Perform pipetting accurately and consistently with these barrier pipette tips in hinged racks, which seal when exposed to potential contaminants. ART hinged racks are available in low retention, micropoint, extended length and other specialty styles.
Unlike standard filter tips, Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks and the covers may be used as hinged or lift-off style. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. ART hinged racks are available in low retention, micropoint, extended length, and other specialty styles.
Features:
  • Stackable hinged racks save space in your lab
  • Sterile tips are certified free from RNase, DNase and pyrogen
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
TRUSTED_SUSTAINABILITY
Specifications
Description ART 10, Filtered, Sterile, Hinged Rack
Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Model ART 10
Length (English) 1.23 in.
Length (Metric) 3.13cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Hinged Rack
Tip Style Universal Fit
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile and PCR Ready