missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARSF Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92826-0.1ml
This item is not returnable.
View return policy
Description
ARSF Polyclonal antibody specifically detects ARSF in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ARSF | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| arylsulfatase F, ASF, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ARSF (NP_004033.2). MRPRRPLVFMSLVCALLNTCQAHRVHDDKPNIVLIMVDDLGIGDLGCYGNDTMRTPHIDRLAREGVRLTQHISAASLCSPSRSAFLTGRYPIRSGMVSSGNRRVIQNLAVPAGLPLNETTLAALLKKQGYSTGLIGKWHQGLNCDSRSDQCHHPYNYGFDYYYGMPFTLVDSCWPDPSRNTELAFESQLWLCVQLVAIAI | |
| 0.1 mL | |
| Cell Biology | |
| 416 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction