missing translation for 'onlineSavingsMsg'
Learn More

Arsenazo II, free acid, >80%, MP Biomedicals™

Product Code. p-4679913 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11470912

Brand: MP Biomedicals 0219505901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 1668-00-4
Molecular Formula C22H18As2N4O14S2
Molecular Weight (g/mol) 776.363
MDL Number MFCD00036695
InChI Key TVMZRHVOFZTNET-RIRMOVKSSA-N
Synonym 2,2'-[1, 8-dihydroxy-3,6-disulfo-2, 7-naphthalene-bis(azo)dibenzenearsonic acid, 2, 2'-[1, 8-dihydroxy-3
PubChem CID 9810878
IUPAC Name (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O

Specifications

Assay Percent Range >80%
Formula Weight 776.37
Quantity 1 g
Physical Form Powder
Chemical Name or Material Arsenazo II, Free Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.