Learn More
ARRY-380 analog, MedChemExpress
Product Code. 16485016
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485016 100 mg 100mg
16484028 10 mg 10mg
16430387 50 mg 50mg
16406747 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16485016 Supplier Medchem Express Supplier No. HY10531
MedChemExpress ARRY-380 analog, an inhibitor of EGFR (ErbB1), is extracted from patent WO2015153959A2, compound 249. ARRY-380 is a potent, selective, ATP-competitive, orally active inhibitor of HER2.
ARRY-380 analog, an inhibitor of EGFR (ErbB1), is extracted from patent WO2015153959A2, compound 249. ARRY-380 is a potent, selective, ATP-competitive, orally active inhibitor of HER2.
Specifications
Chemical Name or Material ARRY-380 analog
CAS 937265-83-3
Color Brown
Purity Grade Notes Research
Molecular Formula C29H27N7O4S
Quantity 100 mg
Solubility Information DMSO : 110 mg/mL (193.11 mM; Need ultrasonic)
SMILES O=S(CCNCC1=CC=C(C2=CC3=C(NC4=CC=C(OC5=CC6=NC=NN6C=C5)C(C)=C4)N=CN=C3C=C2)O1)(C)=O
Molecular Weight (g/mol) 569.63
Formula Weight 569.63
Percent Purity 96.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease