Learn More
Abeba™ Arrow 35600 Shoes
Product Code. 15660188
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. Size unitSize
15660188 44 1 pair
15680178 36 1 pair
15690178 37 1 pair
15600188 38 1 pair
15610188 39 1 pair
15620188 40 1 pair
15630188 41 1 pair
15640188 42 1 pair
15650188 43 1 pair
15670188 45 1 pair
15680188 46 1 pair
15690188 47 1 pair
12 options
Product Code. 15660188
Brand: Abeba™ 35600/44
Only null left
Add to basket
Add to basket
Antistatic with heel patch microfiber. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB A, FO SRA
  • Smooth leather, perforated
  • White
  • Slip resistance PU outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Microfiber heel patch
  • Leather lining
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRA
Color White
Material Leather
Size 44
Sole Features Slip-resistant
Sole Material Polyurethane
Style 35600