Learn More
ARRB1 Monoclonal Antibody (OTI3F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA812547S
Encompass_Preferred
 View more versions of this product
Product Code. 17587024
  • 2816.72 NOK / 30µL
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
ARRB1 Monoclonal antibody specifically detects ARRB1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
ARRB1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P29066, P49407, Q8BWG8
Mouse
Protein A/G
RUO
109689, 25387, 408
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI3F10
Unconjugated
ARRB1
1200006I17Rik; ARB1; ARR1; ARRB1; ARRB1 protein; arrdc1; arrestin; arrestin 2; arrestin beta 1; arrestin beta-1; arrestin, beta 1; arrestin-2; AW208571; BARRES; beta-arrestin; beta-arrestin1; beta-arrestin-1; G430100A01Rik; Non-visual arrestin-2
Full length human recombinant protein of human ARRB1 (NP_004032) produced in E.coli.
30 μL
Primary
Human, Mouse, Rat
Antibody
IgG1