Learn More
ARRB1 Monoclonal Antibody (OTI3F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812547
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
ARRB1 Monoclonal antibody specifically detects ARRB1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ARRB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P29066, P49407, Q8BWG8 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 109689, 25387, 408 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3F10 | |
| Unconjugated | |
| ARRB1 | |
| 1200006I17Rik; ARB1; ARR1; ARRB1; ARRB1 protein; arrdc1; arrestin; arrestin 2; arrestin beta 1; arrestin beta-1; arrestin, beta 1; arrestin-2; AW208571; BARRES; beta-arrestin; beta-arrestin1; beta-arrestin-1; G430100A01Rik; Non-visual arrestin-2 | |
| Full length human recombinant protein of human ARRB1 (NP_004032) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.