Learn More
Thermo Scientific™ ArrayScan™ Liquid Handling Module
Product Code. 15963763 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15963763 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15963763 Supplier Thermo Scientific™ Supplier No. N010117B
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Thermo Scientific™ ArrayScan™ Liquid Handling Module is designed to work with the Thermo Scientific™ ArrayScan™ Live Cell Module, expanding the flexibility of the platform for analysis of complex cellular states of cells.
The ArrayScan Liquid Handling Module has functionalities to aspirate, dispense, and mix, from four deck locations. The module is capable of single-tip aspirate and dispense at any location, creating a highly flexible instrument. Compatible with 6-well, 8-well, 24-well, 96-well, and 384-well plates, the ArrayScan Liquid Handling Module utilizes disposable tips with a dispense volume range of 3 to 200μL.
Features of the ArrayScan Liquid Handling Module:
• Compatible with 6-well, 8-well, 24-well, 96-well, and 384-well plates, the module utilizes disposable tips with a dispense volume range of 3 to 200μL
• Full function liquid handling from any plate or trough (3 plate and 1 quarter trough)
• Industry standard arm with single pipette tip driven by syringe pump (e.g., aspirate, dispense)
• Compatible with all ArrayScan Modules
• Dynamic access to compound and assay plate
• One-to-many and many-to-one transfer scenarios are supported
Includes:
ArrayScan Liquid Handling Module : Automated, single-tip, liquid handling robot module for the Thermo Scientific™ ArrayScan™ High Content Analysis (HCA) Readers using disposable tips (standard and liquid level sensing) to both aspirate and dispense from multiple deck and live cell chamber locations.
TRUSTED_SUSTAINABILITY
Specifications
Product Line ArrayScan
For Use With (Equipment) ArrayScan HCA Readers
Dispense Volume 3 to 200 μL (± 5% in 1 μL increments)
Format 6-wells, 8-wells, 384-well plate, 24-well plate, 96-well plate
Software Compatibility Thermo Scientific™ HCS Studio Cell Analysis Software
Operating Environment Permissible ambient temperature range during operation : + 18 deg C to + 25 deg C, Permissible relative humidity during operation (non-condensing): > 75% (at 30 deg C)
Dimensions 87.63 cm (34.5 in.) (W) x 109.22 cm (43 in.) (H) x 74 cm (29 in.) (D)
Weight (Metric) 59 kg
Quantity 1 each
Voltage 100 V to 240 V
Wattage 150 W (+ 500 W from ArrayScan Reader)