Description
- The Array Card Dye Calibration Kit is used to maintain your real-time PCR instrument with array card block. Dye calibration should be run after lamp change.
- The kit contains FAM™, VIC™, ROX™, ROI, and background calibration plates, as well as FAM™/ROX™ and VIC™/ROX™ normalization plates.
- Please see your instrument user guide for full instructions on use.
Specifications
Specifications
| Calibrated Dye | Proprietary Mix |
| For Use With (Equipment) | ViiA™ 7 Dx System, QuantStudio™ Dx |
| Product Type | Array Card Dye Calibration Kit |
| For Use With (Application) | Real Time PCR (qPCR) |
| Quantity | 7 arrays |
| Calibration Type | Spectral Calibration |
| PCR Method | qPCR |