Learn More
ARPC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16886434 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16886434 150 μL 150µL
16896434 20 μL 20µL
2 options
Product Code. 16886434
Brand: Proteintech 167171AP150UL
Rabbit Polyclonal Antibody
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p16 subunit, has yet to be determined.
Specifications
Antigen ARPC5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ARPC5
Gene Accession No. O15511, Q4KLF8, Q9CPW4
Gene Alias ARC16, ARPC5, p16 Arc
Gene Symbols Arpc5
Host Species Rabbit
Immunogen ARPC5 Fusion Protein Ag10144
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10092, 360854, 67771
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG