missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPC2 Rabbit anti-Human, Mouse, Rat, Clone: 4F2G6, Novus Biologicals™
Rabbit Monoclonal Antibody
1760.00 NOK - 4590.00 NOK
Specifications
| Antigen | ARPC2 |
|---|---|
| Clone | 4F2G6 |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18329216
|
Novus Biologicals
NBP3-15471-20UL |
20 μg |
1760.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18300596
|
Novus Biologicals
NBP3-15471-100UL |
100 μg |
4590.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARPC2 Monoclonal antibody specifically detects ARPC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| ARPC2 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 10109 | |
| IgG | |
| Affinity purified |
| 4F2G6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| actin related protein 2/3 complex, subunit 2 (34 kD), actin related protein 2/3 complex, subunit 2, 34kDa, ARC34actin-related protein 2/3 complex subunit 2, Arp2/3 complex 34 kDa subunit, ARP2/3 protein complex subunit 34, p34-ARC, PNAS-139 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human ARPC2 (O15144). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title