Learn More
ARN272, MedChemExpress
Product Code. 16455266
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16455266
Brand: Medchem Express HY110101
MedChemExpress ARN272 is an anandamide transport inhibitor.
ARN272 is an anandamide transport inhibitor.
Specifications
Chemical Name or Material ARN272
CAS 488793-85-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H20N4O2
Quantity 100 mg
Solubility Information DMSO : 125 mg/mL (289.04 mM; Need ultrasonic)
SMILES OC(C=C1)=CC=C1C2=NN=C(C3=C2C=CC=C3)NC4=CC=C(C(NC5=CC=CC=C5)=O)C=C4
Molecular Weight (g/mol) 432.47
Formula Weight 432.47
Percent Purity 98.38%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.