missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARN-3236, MedChemExpress
Description
ARN-3236 is an oral active and selective inhibitor of salt-inducible kinase 2 (SIK2), with IC50s of <1 nM, 21.63 nM and 6.63 nM for SIK2, SIK1 and SIK3, respectively. Has anti-cancer activity.
Specifications
Specifications
| Chemical Name or Material | ARN-3236 |
| CAS | 1613710-01-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H16N2O2S |
| Quantity | 50 mg |
| Solubility Information | DMSO : 100 mg/mL (297.26 mM; Need ultrasonic) |
| SMILES | COC1=CC=C(C2=CNC3=NC=CC(C4=CSC=C4)=C32)C(OC)=C1 |
| Molecular Weight (g/mol) | 336.41 |
| Formula Weight | 336.41 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?