Learn More
Armenian Hamster IgG Isotype Control (eBio299Arm), PE, eBioscience™, Invitrogen™
Armenian Hamster Isotype Control
950.00 NOK - 2725.00 NOK
Specifications
Antigen | Armenian Hamster IgG |
---|---|
Clone | eBio299Arm |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | PE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15268789
|
Affymetrix eBioscience
12-4888-81 |
50 μg |
950.00 NOK
50µg |
In Stock
Log in to see stock. |
|||||
15476024
|
Affymetrix eBioscience
12-4888-83 |
200 μg |
2725.00 NOK
200µg |
In Stock
Log in to see stock. |
|||||
Description
Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This eBio299Arm antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood. This should be used at the same concentration as the experimental antobody. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.Specifications
Armenian Hamster IgG | |
0.2 mg/mL | |
PE | |
Armenian Hamster | |
Not Applicable | |
IgG; Immunoglobulin G; ImmunoglobulinG | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBio299Arm | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG | |
Affinity chromatography |
For Research Use Only.