missing translation for 'onlineSavingsMsg'
Learn More

ARMCX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20193-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889704

  • 4593.76 NOK / 150µL
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the ALEX family of proteins and may play a role in tumor suppression. The encoded protein contains a potential N-terminal transmembrane domain and two Armadillo (arm) repeats. Other proteins containing the arm repeat are involved in development, maintece of tissue integrity, and tumorigenesis. This gene is closely localized with other family members, including ALEX2 and ALEX3, on the X chromosome.
Specifications

Specifications

ARMCX1
Polyclonal
Unconjugated
ARMCX1
ALEX1, ARMCX1, Protein ALEX1
Rabbit
Antigen Affinity Chromatography
RUO
51309, 78248
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9CX83, Q9P291
ARMCX1
ARMCX1 Fusion Protein Ag14120
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers