missing translation for 'onlineSavingsMsg'
Learn More

ARMC10 Antibody [DyLight 350], Novus Biologicals Biologicals™

Product Code. 30499760 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30499760 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30499760 Supplier Novus Biologicals Supplier No. NBP337916UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARMC10 Polyclonal antibody specifically detects ARMC10 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMC10
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias armadillo repeat containing 10, armadillo repeat-containing protein 10, FLJ37850, MGC3195, specific splicing variant involved in hepatocarcinogenesis, Splicing variant involved in hepatocarcinogenesis protein, SVHPNAS112
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 169-308 of human ARMC10 (NP_001154481.1).,, Sequence:, TNMTVTNDHQHMLHSYITDLFQVLLTGNGNTKVQVLKLLLNLSENPAMTEGLLRAQVDSSFLSLYDSHVAKEILLRVLTLFQNIKNCLKIEGHLAVQPTFTEGSLFFLLHGEECAQKIRALVDHHDAEVKEKVVTIIPKI
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 83787
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.